2,5-Dimethyl-4-hydroxymethylphenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

31152-1G 31152-25G 31152-500MG 31152-5G
76901-926EA 409.51 USD
76901-926 76901-928 76901-930 76901-932
2,5-Dimethyl-4-hydroxymethylphenylboronic acid ≥95%
2,5-Dimethyl-4-hydroxymethylphenylboronic acid
Formula: C₉H₁₃BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD29088269
CAS Number: 2121513-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR