2-Fluoro-3-methylbenzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(2-Fluoro-3-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

31117-25G 31117-5G
76901-904EA 198.39 USD
76901-904 76901-906
2-Fluoro-3-methylbenzeneboronic acid pinacol ester ≥97%
2-Fluoro-3-methylbenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₂
MW: 236.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18384075
CAS Number: 1192548-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR