2-Fluoro-3-methoxypyridine-5-boronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

31073-1G 31073-500MG 31073-5G
76901-870EA 468.92 USD
76901-870 76901-872 76901-874
2-Fluoro-3-methoxypyridine-5-boronic acid pinacol ester ≥97% research grade
2-Fluoro-3-methoxypyridine-5-boronic acid pinacol ester
Formula: C₁₂H₁₇BFNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD16995270
CAS Number: 2121512-67-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR