2-Chloro-3,4-difluorobenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

30938-1G 30938-25G 30938-500MG 30938-5G
76901-780EA 291.75 USD
76901-780 76901-782 76901-784 76901-786
2-Chloro-3,4-difluorobenzaldehyde ≥95%
2-Chloro-3,4-difluorobenzaldehyde
Formula: C₇H₃ClF₂O
Storage Temperature: Refrigerator
MDL Number: MFCD18415551
CAS Number: 1261869-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR