2,6-Dimethyl-4-fluorophenylboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

30902-1G 30902-25G 30902-500MG 30902-5G
76901-764EA 239.77 USD
76901-764 76901-766 76901-768 76901-770
2,6-Dimethyl-4-fluorophenylboronic acid pinacol ester ≥97% research grade
2,6-Dimethyl-4-fluorophenylboronic acid pinacol ester
Formula: C₁₄H₂₀BFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD30189379
CAS Number: 2121512-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR