2,6-Dimethoxy-5-methylpyridine-3-boronic acid 95%

Supplier: AOBChem USA

Warning

30752-1G 30752-500MG 30752-5G 30752-25G
76901-738EA 485.9 USD
76901-738 76901-740 76901-742 77658-596
2,6-Dimethoxy-5-methylpyridine-3-boronic acid 95%
2,6-Dimethoxy-5-methylpyridine-3-boronic acid
Formula: C₈H₁₂BNO₄
Storage Temperature: Refrigerator
MDL Number: MFCD29088194
CAS Number: 2121514-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR