2-Bromo-6-fluoro-3-methylbenzoic acid ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
30716-1G
30716-25G
30716-5G
76901-710EA
77.45
USD
76901-710
76901-712
76901-714
2-Bromo-6-fluoro-3-methylbenzoic acid ≥97% research grade
2-Bromo-6-fluoro-3-methylbenzoic acid
Formula:
C₈H₆BrFO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD22383777 CAS Number: 1359857-60-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...