2-Methoxy-1-naphthoic acid ≥97%

Supplier: AOBChem USA

Warning

30687-25G 30687-5G
76901-698EA 630.18 USD
76901-698 76901-700
2-Methoxy-1-naphthoic acid ≥97%
2-Methoxy-1-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD00021532
CAS Number: 947-62-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR