2-Chloro-6-fluoro-3-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30662-25G 30662-5G
76901-674EA 453.01 USD
76901-674 76901-676
2-Chloro-6-fluoro-3-methylphenylboronic acid pinacol ester ≥97%
2-Chloro-6-fluoro-3-methylphenylboronic acid pinacol ester
Formula: C₁₃H₁₇BClFO₂
MW: 270.535 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29088172
CAS Number: 1688672-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR