4-Fluoro-3-(trifluoromethyl)anisole ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-5-methoxybenzotrifluoride, 1-Fluoro-4-methoxy-2-(trifluoromethyl)benzene

30626-25G 30626-5G
76901-638EA 233.4 USD
76901-638 76901-640
4-Fluoro-3-(trifluoromethyl)anisole ≥95%
4-Fluoro-3-(trifluoromethyl)anisole

Precaución: for Research Use Only

Formula: C₈H₆F₄O
MW: 194.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01631405
CAS Number: 127271-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR