(2-Chloro-5-fluoropyridin-4-yl)methanol ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-5-fluoro-4-(hydroxymethyl)pyridine

30543-1G 30543-25G 30543-500MG 30543-5G
76901-606EA 122.01 USD
76901-606 76901-608 76901-610 76901-612
(2-Chloro-5-fluoropyridin-4-yl)methanol ≥97% research grade
(2-Chloro-5-fluoropyridin-4-yl)methanol
Formula: C₆H₅ClFNO
MW: 161.5600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05662385
CAS Number: 884494-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR