2-Chloro-5-fluoropyridine-4-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

30539-1G 30539-25G 30539-500MG 30539-5G
76901-592EA 150.65 USD
76901-592 76901-594 76901-596 76901-598
2-Chloro-5-fluoropyridine-4-boronic acid pinacol ester ≥97%
2-Chloro-5-fluoropyridine-4-boronic acid pinacol ester
Formula: C₁₁H₁₄BClFNO₂
MW: 257.497 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08063107
CAS Number: 1256360-62-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR