2,6-Dichloro-4-(trifluoromethoxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

30502-1G 30502-25G 30502-5G
76901-568EA 64.72 USD
76901-568 76901-570 76901-572
2,6-Dichloro-4-(trifluoromethoxy)benzoic acid ≥97%
2,6-Dichloro-4-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₃Cl₂F₃O₃
MW: 275.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06660297
CAS Number: 886502-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR