2,6-Difluoro-4-iodophenol ≥97%

Supplier: AOBChem USA

Warning

30432-25G 30432-5G
76901-544EA 364.95 USD
76901-544 76901-546
2,6-Difluoro-4-iodophenol ≥97%
2,6-Difluoro-4-iodophenol

Precaución: for Research Use Only

Formula: C₆H₃F₂IO
Storage Temperature: Ambient
MDL Number: MFCD09907951
CAS Number: 950858-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR