2-Fluoro-4,4'-bipyridine ≥97%

Supplier: AOBChem USA

Warning

30407-1G 30407-500MG 30407-5G
76901-506EA 857.21 USD
76901-506 76901-508 76901-510
2-Fluoro-4,4'-bipyridine ≥97%
2-Fluoro-4,4'-bipyridine
Formula: C₁₀H₇FN₂
MW: 174.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15475748
CAS Number: 1214355-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR