2,6-Dimethoxynicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Dimethoxypyridine-3-carboxylic acid

30349-25G 30349-5G
76901-496EA 477.41 USD
76901-496 76901-498
2,6-Dimethoxynicotinic acid ≥97%
2,6-Dimethoxynicotinic acid
Formula: C₈H₉NO₄
MW: 183.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00075582
CAS Number: 16727-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR