(2-Chloro-3-fluorophenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-3-fluorobenzyl alcohol

30307-25G 30307-5G
76901-488EA 488.02 USD
76901-488 76901-490
(2-Chloro-3-fluorophenyl)methanol ≥97%
(2-Chloro-3-fluorophenyl)methanol
Formula: C₇H₆ClFO
MW: 160.57 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042776
CAS Number: 96516-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR