2-Chloro-5-fluoro-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

30253-1G 30253-25G 30253-5G
76901-470EA 62.6 USD
76901-470 76901-472 76901-474
2-Chloro-5-fluoro-3-methylbenzoic acid ≥97%
2-Chloro-5-fluoro-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆ClFO
Storage Temperature: Ambient
MDL Number: MFCD23712098
CAS Number: 1427416-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR