2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Formyl-4-(trifluoromethyl)phenylboronic acid pinacol ester

30212-1G 30212-25G 30212-500MG 30212-5G
76901-456EA 195.21 USD
76901-456 76901-458 76901-460 76901-462
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)benzaldehyde ≥97%
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-5-(trifluoromethyl)benzaldehyde
Formula: C₁₄H₁₆BF₃O₃
MW: 300.08 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18383807
CAS Number: 1219936-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR