2-Ethoxy-5-fluoroisonicotinic acid ≥97%

Supplier: AOBChem USA

Warning

30059-1G 30059-25G 30059-5G
76901-430EA 102.91 USD
76901-430 76901-432 76901-434
2-Ethoxy-5-fluoroisonicotinic acid ≥97%
2-Ethoxy-5-fluoroisonicotinic acid
Formula: C₈H₈FNO₃
Storage Temperature: Ambient
MDL Number: MFCD28133517
CAS Number: 1809157-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR