2-Chloro-6-(trifluoromethyl)pyridin-4-ol ≥97%
Supplier: AOBChem USA
|
Warning
|
30031-1G
30031-25G
30031-500MG
30031-5G
76901-414EA
409.51
USD
76901-414
76901-416
76901-418
76901-420
2-Chloro-6-(trifluoromethyl)pyridin-4-ol ≥97%
2-Chloro-6-(trifluoromethyl)pyridin-4-ol
Formula:
C₆H₃ClF₃NO MW: 197.542 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16610509 CAS Number: 1227602-42-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...