[2-Chloro-6-(trifluoromethyl)pyridin-4-yl]boronic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
(2-Chloro-6-(trifluoromethyl)pyridin-4-yl)boronic acid, 2-Chloro-6-(trifluoromethyl)pyridine-4-boronic acid
30028-1G
30028-25G
30028-500MG
30028-5G
76901-398EA
332.07
USD
76901-398
76901-400
76901-402
76901-404
[2-Chloro-6-(trifluoromethyl)pyridin-4-yl]boronic acid ≥97%
[2-Chloro-6-(trifluoromethyl)pyridin-4-yl]boronic acid
Precaución: for Research Use Only
Formula:
C₆H₄BClF₃NO₂ MW: 225.361 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD13188845 CAS Number: 1446486-10-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...