2-Chloro-4-hydroxymethyl-3-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

29916-1G 29916-25G 29916-500MG 29916-5G
76901-374EA 242.95 USD
76901-374 76901-376 76901-378 76901-380
2-Chloro-4-hydroxymethyl-3-methylphenylboronic acid ≥97%
2-Chloro-4-hydroxymethyl-3-methylphenylboronic acid
Formula: C₈H₁₀BClO₃
MW: 200.427 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD29036612
CAS Number: 2121512-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR