2-Chloro-3-methoxy-6-(trifluoromethyl)pyridine ≥97% research grade

Supplier: AOBChem USA

Warning

29839-1G 29839-25G 29839-500MG 29839-5G
76901-358EA 162.32 USD
76901-358 76901-360 76901-362 76901-364
2-Chloro-3-methoxy-6-(trifluoromethyl)pyridine ≥97% research grade
2-Chloro-3-methoxy-6-(trifluoromethyl)pyridine
Formula: C₇H₅ClF₃NO
Storage Temperature: Refrigerator
MDL Number: MFCD15478057
CAS Number: 1214348-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR