2-Chloroisonicotinoyl chloride ≥97% research grade

Supplier: AOBChem USA

Warning

29719-25G 29719-5G
76901-334EA 559.1 USD
76901-334 76901-336
2-Chloroisonicotinoyl chloride ≥97% research grade
2-Chloroisonicotinoyl chloride
Formula: C₆H₃Cl₂NO
Storage Temperature: Refrigerator
MDL Number: MFCD00204232
CAS Number: 65287-34-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR