2-Bromo-4,6-dichlorophenol ≥97%

Supplier: AOBChem USA

Warning

29633-25G 29633-5G
76901-322EA 630.18 USD
76901-322 76901-324
2-Bromo-4,6-dichlorophenol ≥97%
2-Bromo-4,6-dichlorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrCl₂O
Storage Temperature: Ambient
CAS Number: 4524-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR