2-Bromo-3,4-difluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

29585-25G 29585-5G
76901-312EA 230.22 USD
76901-312 76901-314
2-Bromo-3,4-difluorobenzoic acid ≥97%
2-Bromo-3,4-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O₂
MW: 236.998 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09835210
CAS Number: 170108-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR