2-Bromo-3,4-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

29583-1G 29583-25G 29583-5G
76901-298EA 96.55 USD
76901-298 76901-300 76901-302
2-Bromo-3,4-difluorobenzaldehyde ≥97%
2-Bromo-3,4-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O
MW: 220.999 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28669844
CAS Number: 1578156-21-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR