2-Chloro-4,5-difluorophenylboronic acid, pinacol ester ≥97%

Supplier: AOBChem USA

Warning

29474-1G 29474-25G 29474-500MG 29474-5G
76901-260EA 357.53 USD
76901-260 76901-262 76901-264 76901-266
2-Chloro-4,5-difluorophenylboronic acid, pinacol ester ≥97%
2-Chloro-4,5-difluorophenylboronic acid, pinacol ester
Formula: C₁₂H₁₄BClF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD28806036
CAS Number: 2121514-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR