2-Bromo-3-isopropoxypyridine ≥97%

Supplier: AOBChem USA

Warning

29388-1G 29388-25G 29388-500MG 29388-5G
76901-244EA 127.31 USD
76901-244 76901-246 76901-248 76901-250
2-Bromo-3-isopropoxypyridine ≥97%
2-Bromo-3-isopropoxypyridine
Formula: C₈H₁₀BrNO
Storage Temperature: Ambient
MDL Number: MFCD11878542
CAS Number: 113503-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR