2-Isopropyl-4-methoxyphenyl boronic acid ≥97%

Supplier: AOBChem USA

Warning

29330-1G 29330-500MG 29330-5G
76901-230EA 831.75 USD
76901-230 76901-232 76901-234
2-Isopropyl-4-methoxyphenyl boronic acid ≥97%
2-Isopropyl-4-methoxyphenyl boronic acid
Formula: C₁₀H₁₅BO₃
MW: 194.035 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27949212
CAS Number: 313667-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR