2-Ethoxy-6-fluoro-4-methylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

29318-1G 29318-25G 29318-500MG 29318-5G
76901-214EA 225.98 USD
76901-214 76901-216 76901-218 76901-220
2-Ethoxy-6-fluoro-4-methylphenylboronic acid ≥97%
2-Ethoxy-6-fluoro-4-methylphenylboronic acid
Formula: C₉H₁₂BFO₃
MW: 197.999 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28806033
CAS Number: 2121513-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR