2,4-Difluoro-5-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

29292-1G 29292-25G 29292-500MG 29292-5G
76901-192EA 290.69 USD
76901-192 76901-194 76901-196 76901-198
2,4-Difluoro-5-formylphenylboronic acid ≥97%
2,4-Difluoro-5-formylphenylboronic acid
Formula: C₇H₅BF₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD28147737
CAS Number: 1413393-42-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR