2-Bromo-4-iodophenol 95%

Supplier: AOBChem USA

Warning

29282-1G 29282-25G 29282-5G 29282-500MG
76901-182EA 106.09 USD
76901-182 76901-184 76901-186 77645-800
2-Bromo-4-iodophenol 95%
2-Bromo-4-iodophenol
Formula: C₆H₄BrIO
MW: 298.9038 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11870189
CAS Number: 133430-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR