2-Methyl-4-(methylthio)benzoic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Methyl-4-(methylsulphanyl)benzoic acid

29139-25G 29139-5G
76901-144EA 325.7 USD
76901-144 76901-146
2-Methyl-4-(methylthio)benzoic acid ≥97% research grade
2-Methyl-4-(methylthio)benzoic acid
Formula: C₉H₁₀O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD20651197
CAS Number: 118939-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR