2-Hydroxy-4-methoxy phenyl boronic acid pinacol ester ≥97% research grade
Supplier: AOBChem USA
|
Warning
|
29094-1G
29094-25G
29094-500MG
29094-5G
76901-128EA
290.69
USD
76901-128
76901-130
76901-132
76901-134
2-Hydroxy-4-methoxy phenyl boronic acid pinacol ester ≥97% research grade
2-Hydroxy-4-methoxy phenyl boronic acid pinacol ester
Formula:
C₁₃H₁₉BO₄ MW: 250.099 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16994338 CAS Number: 2072801-99-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...