2-Bromo-3-fluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Bromo-3-fluorophenyl)methanol

29091-1G 29091-25G 29091-5G
76901-116EA 112.46 USD
76901-116 76901-118 76901-120
2-Bromo-3-fluorobenzyl alcohol ≥97%
2-Bromo-3-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₆BrFO
MW: 205.024 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12911577
CAS Number: 1184915-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR