2-Chloro-5-iodobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

29062-1G 29062-500MG 29062-5G 29062-25G
76901-106EA 464.68 USD
76901-106 76901-108 76901-110 77658-500
2-Chloro-5-iodobenzaldehyde ≥97%
2-Chloro-5-iodobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄ClIO
Storage Temperature: Refrigerator
MDL Number: MFCD12024988
CAS Number: 256925-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR