2-Fluoro-3-hydroxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

28797-1G 28797-25G 28797-500MG 28797-5G
76901-070EA 274.78 USD
76901-070 76901-072 76901-074 76901-076
2-Fluoro-3-hydroxyphenylboronic acid pinacol ester ≥97%
2-Fluoro-3-hydroxyphenylboronic acid pinacol ester
Formula: C₁₂H₁₆BFO₃
MW: 238.063 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16994264
CAS Number: 2121514-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR