2',5'-Dibromo-4'-methoxyacetophenone ≥95%

Supplier: AOBChem USA

Warning

28745-25G 28745-5G
76901-058EA 453.01 USD
76901-058 76901-060
2',5'-Dibromo-4'-methoxyacetophenone ≥95%
2',5'-Dibromo-4'-methoxyacetophenone
Formula: C₉H₈Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD28360706
CAS Number: 1809168-60-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR