ω-Chloro-ω,ω-difluoroacetophenone ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-2,2-difluoro-1-phenylethanone, α-Chloro-α,α-difluoroacetophenone, 2-Chloro-2,2-difluoroacetophenone

28681-25G 28681-5G
76901-032EA 286.45 USD
76901-032 76901-034
ω-Chloro-ω,ω-difluoroacetophenone ≥97%
ω-Chloro-ω,ω-difluoroacetophenone

Precaución: for Research Use Only

Formula: (C₆H₅)C(O)CClF₂
MW: 190.58 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00042123
CAS Number: 384-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR