2-Bromo-5-phenylthiophene ≥95%

Supplier: AOBChem USA

Warning

28573-25G 28573-5G
76901-022EA 233.4 USD
76901-022 76901-024
2-Bromo-5-phenylthiophene ≥95%
2-Bromo-5-phenylthiophene

Precaución: for Research Use Only

Formula: C₁₀H₇BrS
MW: 239.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00159575
CAS Number: 29488-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR