2-Iodo-1-methylimidazole ≥97%

Supplier: AOBChem USA

Warning

28422-25G 28422-5G
76900-990EA 587.74 USD
76900-990 76900-992
2-Iodo-1-methylimidazole ≥97%
2-Iodo-1-methylimidazole

Precaución: for Research Use Only

Formula: C₄H₅IN₂
Storage Temperature: Ambient
MDL Number: MFCD00661606
CAS Number: 37067-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR