2,5-Dimethyl-4-formylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

28331-1G 28331-25G 28331-500MG 28331-5G
76900-950EA 231.28 USD
76900-950 76900-952 76900-954 76900-956
2,5-Dimethyl-4-formylphenylboronic acid pinacol ester ≥97%
2,5-Dimethyl-4-formylphenylboronic acid pinacol ester
Formula: C₁₅H₂₁BO₃
Storage Temperature: Ambient
MDL Number: MFCD18383424
CAS Number: 1256970-23-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR