2-Cyano-5-fluorophenylboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (2-Cyano-5-fluorophenyl)boronic acid

28243-1G 28243-25G 28243-5G
76900-926EA 115.64 USD
76900-926 76900-928 76900-930
2-Cyano-5-fluorophenylboronic acid ≥95%
2-Cyano-5-fluorophenylboronic acid
Formula: C₇H₅BFNO₂
MW: 164.93 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18394078
CAS Number: 1375109-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR