2-Hydroxy-3-(trifluoromethyl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 3-(Trifluoromethyl)salicylaldehyde

28195-1G 28195-25G 28195-5G
76900-914EA 63.66 USD
76900-914 76900-916 76900-918
2-Hydroxy-3-(trifluoromethyl)benzaldehyde ≥97%
2-Hydroxy-3-(trifluoromethyl)benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03788529
CAS Number: 336628-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR