1,3-Dibromo-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

28107-25G 28107-5G
76900-892EA 318.27 USD
76900-892 76900-894
1,3-Dibromo-2-iodobenzene ≥97%
1,3-Dibromo-2-iodobenzene
Formula: C₆H₃Br₂I
Storage Temperature: Ambient
MDL Number: MFCD00192666
CAS Number: 19821-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR