(2,6-Dibromopyridin-4-yl)methanol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Dibromo-4-hydroxymethylpyridine

28033-1G 28033-25G 28033-5G
76900-880EA 67.9 USD
76900-880 76900-882 76900-884
(2,6-Dibromopyridin-4-yl)methanol ≥97%
(2,6-Dibromopyridin-4-yl)methanol
Formula: C₆H₅Br₂NO
MW: 266.91 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09836667
CAS Number: 223463-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR