2-Bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromothiazole-5-boronic acid pinacol ester

27973-1G 27973-25G 27973-500MG 27973-5G
76900-864EA 332.07 USD
76900-864 76900-866 76900-868 76900-870
2-Bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiazole ≥97%
2-Bromo-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)thiazole
Formula: C₉H₁₃BBrNO₂S
MW: 289.98 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13181957
CAS Number: 1402166-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR