2,6-Dimethyl-4-formyl-benzoic acid ≥97%

Supplier: AOBChem USA

Warning

27870-1G 27870-25G 27870-500MG 27870-5G
76900-834EA 201.58 USD
76900-834 76900-836 76900-838 76900-840
2,6-Dimethyl-4-formyl-benzoic acid ≥97%
2,6-Dimethyl-4-formyl-benzoic acid
Formula: C₁₀H₁₀O₃
MW: 178.185 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20645598
CAS Number: 306296-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR